Assay Details
Target Gene Details
Entrez Gene ID: | 18193 |
Gene Name: | nuclear receptor-binding SET-domain protein 1 |
Gene Aliases: |
AI528500, KMT3B |
Location: |
Chr.13:55209782-55318325 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsd1 | NM_008739.3 | NP_032765.3 | ||
CB247038.1 | ||||
CD806124.1 |
