Assay Details
Target Gene Details
Entrez Gene ID: | 18193 |
Gene Name: | nuclear receptor-binding SET-domain protein 1 |
Gene Aliases: |
AI528500, KMT3B |
Location: |
Chr.13:55209782-55318325 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsd1 | NM_008739.3 | 4 | 1260 | NP_032765.3 |
AF419220.1 | 2 | 219 | AAL40940.1 | |
AK145783.1 | 4 | 1261 | BAE26648.1 | |
AK164087.1 | 5 | 456 | ||
AK172676.1 | 5 | 754 | ||
BB687537.1 | 3 | 357 | ||
BY389912.1 | 1 | 94 |
