Assay Details
Target Gene Details
Entrez Gene ID: | 435350 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6e |
Gene Aliases: |
Gm11396, OTTMUSG00000000720, SPI3B, ovalbumin |
Location: |
Chr.13:33832292-33843408 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb6e | NM_001045535.2 | NP_001039000.2 |
More Information
Set Membership: |
|

Genomic Map