Assay Details
Target Gene Details
Entrez Gene ID: | 218138 |
Gene Name: | GDP-mannose 4, 6-dehydratase |
Gene Aliases: |
BC031788, C87208 |
Location: |
Chr.13:31819583-32338725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmds | NM_146041.2 | NP_666153.1 | ||
| BC031788.1 | AAH31788.1 | |||
| BC093502.1 | AAH93502.1 |
More Information
Set Membership: |
|

Genomic Map