Assay Details
Target Gene Details
Entrez Gene ID: | 218203 |
Gene Name: | myosin regulatory light chain interacting protein |
Gene Aliases: |
9430057C20Rik, Mir |
Location: |
Chr.13:45389742-45411940 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mylip | NM_153789.3 | NP_722484.2 | ||
| AK034887.1 | BAC28868.1 | |||
| AK040836.1 | BAC30716.1 | |||
| AK159478.1 | BAE35115.1 | |||
| AK171888.1 | BAE42721.1 | |||
| AK193807.1 | ||||
| AK202678.1 | ||||
| AY434449.1 | AAQ98867.1 | |||
| BC010206.1 | AAH10206.1 |

Genomic Map