Assay Details
Target Gene Details
Entrez Gene ID: | 218630 |
Gene Name: | cyclin O |
Gene Aliases: |
C86987, Ccnu, E130318K11, Ung2 |
Location: |
Chr.13:112987802-112990778 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccno | NM_001081062.1 | 2 | 530 | NP_001074531.1 |
| AK053890.1 | 1 | 1135 | ||
| AK086507.1 | 2 | 505 | ||
| BC147759.1 | 2 | 506 | ||
| BC147766.1 | 2 | 506 |
More Information
Set Membership: |
|

Genomic Map