Assay Details
Target Gene Details
Entrez Gene ID: | 72935 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 41 |
Gene Aliases: |
2900024F02Rik, AA958953, ABS, AI324246 |
Location: |
Chr.13:55530410-55536658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx41 | NM_134059.2 | NP_598820.2 | ||
AK013586.1 | BAB28917.1 | |||
AK151427.1 | BAE30391.1 | |||
AK156957.1 | BAE33914.1 | |||
AK168043.1 | BAE40025.1 | |||
BC011308.1 | AAH11308.1 |
