Assay Details
Target Gene Details
Entrez Gene ID: | 212880 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 46 |
Gene Aliases: |
2200005K02Rik, 8430438J23Rik, AI325430, AI957095, mKIAA0801 |
Location: |
Chr.13:55634969-55681267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx46 | NM_001282055.1 | NP_001268984.1 | ||
AK088950.1 | 12 | 3352 | ||
AK129220.1 | 12 | 3353 | BAC98030.2 | |
AK134073.1 | 1 | 1008 | BAE22002.1 | |
BC026492.1 | AAH26492.1 | |||
BC092240.1 | AAH92240.1 |
