Assay Details
Target Gene Details
Entrez Gene ID: | 78038 |
Gene Name: | methylcrotonoyl-Coenzyme A carboxylase 2 (beta) |
Gene Aliases: |
4930552N12Rik, MCCB |
Location: |
Chr.13:99948532-100015639 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mccc2 | NM_030026.2 | NP_084302.1 | ||
| AK019760.1 | 7 | 573 | BAB31847.1 | |
| AK132265.1 | BAE21068.1 | |||
| AK143233.1 | BAE25319.1 | |||
| AK145564.1 | BAE26513.1 | |||
| AK146844.1 | BAE27475.1 | |||
| AK146865.1 | BAE27489.1 | |||
| BC027640.1 | AAH27640.1 | |||
| BC058987.1 | AAH58987.1 | |||
| BC141193.1 | ||||
| BC141194.1 |
More Information
Set Membership: |
|

Genomic Map