Assay Details
Target Gene Details
Entrez Gene ID: | 14945 |
Gene Name: | granzyme K |
Gene Aliases: |
- |
Location: |
Chr.13:113171874-113180897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzmk | NM_008196.1 | NP_032222.1 | ||
| BC117981.1 |
More Information
Set Membership: |
|

Genomic Map