Assay Details
Target Gene Details
Entrez Gene ID: | 238896 |
Gene Name: | cell division cycle 20B |
Gene Aliases: |
EG238896, EG622422 |
Location: |
Chr.13:113032954-113121019 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc20b | NM_001281487.1 | NP_001268416.1 |
More Information
Set Membership: |
|

Genomic Map