Assay Details
Target Gene Details
Entrez Gene ID: | 73274 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
1700034P14Rik, AU019836, D230035M11Rik, Gpbp, mGPBP |
Location: |
Chr.13:111425680-111490123 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpbp1 | NM_001122963.1 | 4 | 1455 | NP_001116435.1 |
| NM_028487.4 | 4 | 1455 | NP_082763.3 | |
| AK082704.1 | 4 | 1273 | BAC38578.1 | |
| AK135728.1 | 4 | 1386 | BAE22631.1 | |
| AK136894.1 | 4 | 1234 | ||
| AK145318.1 | 4 | 1444 | BAE26364.1 | |
| AK151283.1 | 3 | 676 | ||
| AK161928.1 | 4 | 1243 | BAE36638.1 | |
| AK163370.1 | 4 | 1303 | BAE37320.1 | |
| AK163390.1 | 4 | 1457 | BAE37330.1 | |
| AK169406.1 | 4 | 1250 | BAE41152.1 | |
| AY382529.1 | 4 | 1253 | AAQ88446.1 | |
| BC016083.1 | 3 | 473 | AAH16083.1 | |
| BC067075.1 | 4 | 1229 | AAH67075.1 | |
| BC094888.1 | 4 | 1248 | AAH94888.1 |

Genomic Map