Assay Details
Target Gene Details
Entrez Gene ID: | 73274 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
1700034P14Rik, AU019836, D230035M11Rik, Gpbp, mGPBP |
Location: |
Chr.13:111425680-111490123 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpbp1 | NM_001122963.1 | NP_001116435.1 | ||
| NM_028487.4 | NP_082763.3 | |||
| AK082704.1 | BAC38578.1 | |||
| AK135728.1 | BAE22631.1 | |||
| AK136894.1 | ||||
| AK145318.1 | BAE26364.1 | |||
| AK151283.1 | ||||
| AK161928.1 | BAE36638.1 | |||
| AK163370.1 | BAE37320.1 | |||
| AK163390.1 | BAE37330.1 | |||
| AK169406.1 | BAE41152.1 | |||
| AY382529.1 | AAQ88446.1 | |||
| BC016083.1 | AAH16083.1 | |||
| BC067075.1 | AAH67075.1 | |||
| BC094888.1 | AAH94888.1 |

Genomic Map