Assay Details
Target Gene Details
Entrez Gene ID: | 26914 |
Gene Name: | H2A histone family, member Y |
Gene Aliases: |
H2AF12M, mH2a1, macroH2A1 |
Location: |
Chr.13:56073622-56135626 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2afy | NM_001159513.1 | NP_001152985.1 | ||
NM_001159514.1 | NP_001152986.1 | |||
NM_001159515.1 | NP_001152987.1 | |||
NM_012015.2 | NP_036145.1 | |||
AB071988.2 | BAB68541.1 | |||
AF171080.1 | AAD53745.1 | |||
AK039274.1 | BAC30302.1 | |||
AK177545.1 | ||||
AK177665.1 | ||||
AK180236.1 | ||||
AK182998.1 | ||||
AK184583.1 | ||||
AK185330.1 | ||||
AK186090.1 | ||||
AK188813.1 | ||||
AK192900.1 | ||||
AK193982.1 | ||||
AK199563.1 | ||||
AK201036.1 | ||||
AK208400.1 | ||||
AK209231.1 | ||||
AK213934.1 | ||||
AW990415.1 | ||||
BC006955.1 | AAH06955.1 | |||
CB574813.1 | ||||
CV561613.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
