Assay Details
Target Gene Details
Entrez Gene ID: | 13206 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 4 |
Gene Aliases: |
AV206478, Mvh, VASA |
Location: |
Chr.13:112598333-112652629 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx4 | NM_001145885.1 | 16 | 1287 | NP_001139357.1 |
NM_010029.2 | 15 | 1209 | NP_034159.1 | |
AK014844.1 | 15 | 1200 | BAB29578.1 | |
AK133366.1 | 16 | 1280 | BAE21618.1 | |
BC137601.1 | 15 | 1263 | ||
BC144760.1 | 15 | 1263 | ||
D14859.1 | 12 | 982 | BAA03584.1 | |
JQ923486.1 | 15 | 1237 |
