Assay Details
Target Gene Details
Entrez Gene ID: | 19111 |
Gene Name: | prolactin family 6, subfamily a, member 1 |
Gene Aliases: |
PLP-B, Prlpb |
Location: |
Chr.13:27312627-27319252 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl6a1 | NM_011166.2 | 2 | 150 | NP_035296.1 |
| AF011384.1 | 2 | 149 | AAB92400.1 | |
| AF015563.1 | 2 | 150 | AAB68825.1 |
More Information
Set Membership: |
|

Genomic Map