Assay Details
Target Gene Details
Entrez Gene ID: | 12759 |
Gene Name: | clusterin |
Gene Aliases: |
AI893575, ApoJ, Cli, D14Ucla3, SP-40, Sgp-2, Sgp2, Sugp-2 |
Location: |
Chr.14:65968483-65981547 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clu | NM_013492.2 | 4 | 589 | NP_038520.2 |
AF248057.1 | 4 | 472 | AAF67184.1 | |
AF248058.1 | 1 | 143 | AAF67185.1 | |
BC075668.1 | 4 | 418 | AAH75668.1 | |
D14077.1 | 4 | 447 | BAA03162.1 | |
L05670.1 | 4 | 447 | AAA37284.1 | |
L08235.1 | 3 | 419 | AAA37422.1 | |
S70244.1 | 3 | 383 | AAB30623.1 |
