Assay Details
Target Gene Details
Entrez Gene ID: | 69716 |
Gene Name: | thyroid hormone receptor interactor 13 |
Gene Aliases: |
2410002G23Rik, D13Ertd328e |
Location: |
Chr.13:73912462-73937767 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip13 | NM_027182.2 | NP_081458.1 | ||
| AK010336.1 | BAB26861.1 | |||
| AK146877.1 | BAE27499.1 | |||
| AK151568.1 | BAE30510.1 | |||
| AK212754.1 | ||||
| AK215913.1 | ||||
| BC023834.1 | ||||
| BC126946.1 |
More Information
Set Membership: |
|

Genomic Map