Assay Details
Target Gene Details
Entrez Gene ID: | 73244 |
Gene Name: | prolactin family 8, subfamily a, member 1 |
Gene Aliases: |
1600017L04Rik, 3830403L08Rik, PLP-Cd, Plpcd, Prlpc4 |
Location: |
Chr.13:27573922-27582171 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl8a1 | NM_028477.2 | NP_082753.1 | ||
| AF525158.1 | AAN39706.1 | |||
| AK005485.1 | BAB24071.1 | |||
| AK014414.1 | BAB29336.1 | |||
| BC109185.1 | AAI09186.1 | |||
| BC109186.1 | AAI09187.1 |
More Information
Set Membership: |
|

Genomic Map