Assay Details
Target Gene Details
Entrez Gene ID: | 18676 |
Gene Name: | PHD finger protein 2 |
Gene Aliases: |
GRC5 |
Location: |
Chr.13:48801750-48870885 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf2 | NM_011078.3 | NP_035208.2 | ||
AB221591.1 | ||||
AF043726.1 | AAD21792.1 | |||
AK153906.1 | ||||
AK172995.1 | BAD32273.1 | |||
BC051633.1 | AAH51633.1 |
