Assay Details
Target Gene Details
Entrez Gene ID: | 19112 |
Gene Name: | prolactin family 8, subfamily a, member 6 |
Gene Aliases: |
PLP-C1, PLP-Ca, PLP-calpha, Prlpc, Prlpc1 |
Location: |
Chr.13:27432681-27438688 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl8a6 | NM_001271378.1 | NP_001258307.1 | ||
| NM_001271379.1 | NP_001258308.1 | |||
| NM_011167.3 | NP_035297.1 | |||
| NR_073166.1 | ||||
| AF090140.1 | AAD09012.1 | |||
| AK005435.1 | BAB24029.1 | |||
| AK005478.1 | BAB24067.1 | |||
| BC099469.1 | AAH99469.1 |
More Information
Set Membership: |
|

Genomic Map