Assay Details
Target Gene Details
Entrez Gene ID: | 67288 |
Gene Name: | splicing regulatory glutamine/lysine-rich protein 1interacting protein 1 |
Gene Aliases: |
3110031B13Rik, AI462702, AV348305, AW121048, Sfrs12ip1, Srsf12ip1 |
Location: |
Chr.13:104817239-104838659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srek1ip1 | NM_026075.2 | NP_080351.2 | ||
| AK013737.1 | ||||
| AK014102.1 | BAB29157.1 | |||
| AK018357.1 | BAB31175.1 | |||
| AK020113.1 | BAB31999.3 | |||
| BC027133.1 | ||||
| BC038302.1 | AAH38302.1 | |||
| BC096661.1 | AAH96661.1 | |||
| BC116993.1 | ||||
| BC116999.1 |

Genomic Map