Assay Details
Target Gene Details
Entrez Gene ID: | 268706 |
Gene Name: | solute carrier family 38, member 9 |
Gene Aliases: |
4833412L08Rik, 6720411P22Rik, 9130023D20Rik, 9430067K09Rik, A730092C09 |
Location: |
Chr.13:112660691-112738752 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc38a9 | NM_178746.4 | NP_848861.1 | ||
| AK043402.1 | BAC31537.1 | |||
| AK044735.1 | BAC32057.1 | |||
| AK078926.1 | BAC37465.1 | |||
| BC052361.1 | AAH52361.1 |
More Information
Set Membership: |
|

Genomic Map