Assay Details
Target Gene Details
Entrez Gene ID: | 21371 |
Gene Name: | tubulin cofactor A |
Gene Aliases: |
Tbca13 |
Location: |
Chr.13:94788943-94842899 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbca | NM_009321.2 | NP_033347.1 | ||
| AK010859.1 | BAB27228.1 | |||
| AK140276.1 | BAE24312.1 | |||
| AK151218.1 | BAE30212.1 | |||
| AK168296.1 | BAE40239.1 | |||
| BC034267.1 | AAH34267.1 | |||
| BC051475.1 | AAH51475.1 | |||
| BC055749.1 | AAH55749.1 | |||
| BC087931.1 | AAH87931.1 | |||
| U05333.1 | AAA83250.1 |
More Information
Set Membership: |
|

Genomic Map