Assay Details
Target Gene Details
Entrez Gene ID: | 218460 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
B830029I03Rik, MSTP048 |
Location: |
Chr.13:94976213-95023316 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wdr41 | NM_172590.3 | 1 | 122 | NP_766178.2 |
AK046853.1 | 1 | 91 | BAC32897.1 | |
AK082348.1 | 1 | 123 | BAC38474.1 | |
AK149993.1 | 1 | 88 | BAE29221.1 | |
BB650073.1 | 1 | 118 | ||
BC138095.1 | ||||
BC145144.1 |
