Assay Details
Target Gene Details
Entrez Gene ID: | 105518 |
Gene Name: | RIKEN cDNA A630023A22 gene |
Gene Aliases: |
AI461879 |
Location: |
Chr.14:34051126-34102754 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630023A22Rik | NM_001251843.1 | NP_001238772.1 | ||
| AK041587.1 | ||||
| AW989347.1 |
Target Gene Details
Entrez Gene ID: | 239029 |
Gene Name: | anthrax toxin receptor-like |
Gene Aliases: |
1700112N15Rik, 4933430J11 |
Location: |
Chr.14:34052713-34076323 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Antxrl | NM_172808.2 | NP_766396.2 | ||
| AK077206.1 | BAC36683.1 |
More Information
Set Membership: |
|

Genomic Map