Assay Details
Target Gene Details
Entrez Gene ID: | 54373 |
Gene Name: | protease, serine 16 (thymus) |
Gene Aliases: |
AI448615, TSSP |
Location: |
Chr.13:22002176-22009741 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss16 | NM_019429.2 | NP_062302.1 | ||
| AJ131775.1 | CAB66137.1 | |||
| AK037514.1 | ||||
| AK088019.1 | 1 | 85 | BAC40100.1 | |
| BC125481.1 | ||||
| BC131955.1 |
More Information
Set Membership: |
|

Genomic Map