Assay Details
Target Gene Details
Entrez Gene ID: | 108912 |
Gene Name: | cell division cycle associated 2 |
Gene Aliases: |
2610311M19Rik, AI586158 |
Location: |
Chr.14:67673180-67715841 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca2 | NM_001110162.1 | 10 | 1609 | NP_001103632.1 |
NM_175384.4 | 10 | 1312 | NP_780593.3 | |
AK031313.1 | 6 | 754 | BAC27343.1 | |
AK160223.1 | 10 | 1371 | ||
AK166952.1 | 10 | 1379 | ||
BC116298.1 | 6 | 754 | ||
BC116299.1 | 6 | 754 |
