Assay Details
Target Gene Details
Entrez Gene ID: | 108912 |
Gene Name: | cell division cycle associated 2 |
Gene Aliases: |
2610311M19Rik, AI586158 |
Location: |
Chr.14:67673180-67715841 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca2 | NM_001110162.1 | 6 | 1008 | NP_001103632.1 |
NM_175384.4 | 6 | 711 | NP_780593.3 | |
AK031083.1 | 6 | 774 | BAC27245.1 | |
AK031313.1 | 2 | 153 | BAC27343.1 | |
AK160223.1 | 6 | 770 | ||
AK166952.1 | 6 | 777 | ||
BC116298.1 | 2 | 153 | ||
BC116299.1 | 2 | 153 |
