Assay Details
Target Gene Details
Entrez Gene ID: | 69666 |
Gene Name: | proteasome (prosome, macropain) assembly chaperone 4 |
Gene Aliases: |
PAC-4, mPAC4 |
Location: |
Chr.13:34162964-34178172 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmg4 | NM_001101430.2 | NP_001094900.1 | ||
| NM_001110515.2 | NP_001103985.1 | |||
| AA063946.1 | ||||
| AA124813.1 | ||||
| AK009887.1 |
More Information
Set Membership: |
|

Genomic Map