Assay Details
Target Gene Details
Entrez Gene ID: | 56196 |
Gene Name: | tyrosyl-DNA phosphodiesterase 2 |
Gene Aliases: |
D13Ertd656e, Ttrap |
Location: |
Chr.13:24831647-24842153 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdp2 | NM_019551.2 | NP_062424.1 | ||
| AJ251328.1 | CAB92971.1 | |||
| AK009089.1 | ||||
| AK135925.1 | ||||
| AK167707.1 | BAE39751.1 | |||
| AK180792.1 | ||||
| BC132511.1 | ||||
| BC132513.1 | ||||
| BY719852.1 |

Genomic Map