Assay Details
Target Gene Details
Entrez Gene ID: | 56196 |
Gene Name: | tyrosyl-DNA phosphodiesterase 2 |
Gene Aliases: |
D13Ertd656e, Ttrap |
Location: |
Chr.13:24831647-24842153 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tdp2 | NM_019551.2 | NP_062424.1 | ||
AJ251328.1 | CAB92971.1 | |||
AK009089.1 | ||||
AK135925.1 | ||||
AK167707.1 | BAE39751.1 | |||
AK180792.1 | ||||
BC132511.1 | ||||
BC132513.1 |
