Assay Details
Target Gene Details
Entrez Gene ID: | 18740 |
Gene Name: | paired-like homeodomain transcription factor 1 |
Gene Aliases: |
Bft, P-OTX, Potx, Ptx1 |
Location: |
Chr.13:55825046-55836194 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx1 | NM_011097.2 | NP_035227.1 | ||
| AK142455.1 | BAE25071.1 | |||
| BC012696.1 | AAH12696.1 | |||
| U54499.1 | AAC52716.1 | |||
| U71206.1 | AAB16860.1 |
More Information
Set Membership: |
|

Genomic Map