Assay Details
Target Gene Details
Entrez Gene ID: | 22151 |
Gene Name: | tubulin, beta 2A class IIA |
Gene Aliases: |
M(beta)2, Tubb2 |
Location: |
Chr.13:34074278-34078008 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb2a | NM_009450.2 | NP_033476.1 | ||
| AK134545.1 | BAE22178.1 | |||
| BC055441.1 | AAH55441.1 | |||
| CJ148662.1 |
More Information
Set Membership: |
|

Genomic Map