Assay Details
Target Gene Details
Entrez Gene ID: | 69672 |
Gene Name: | thioredoxin domain containing 15 |
Gene Aliases: |
2310047H23Rik, AI854086, C5orf14, UNQ335 |
Location: |
Chr.13:55714650-55726227 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Txndc15 | NM_175150.3 | NP_780359.2 | ||
AK009878.1 | ||||
AK079517.1 | BAC37669.1 | |||
AK160051.1 | BAE35591.1 | |||
AK167701.1 | BAE39745.1 | |||
AK167838.1 | BAE39859.1 | |||
AK167977.1 | BAE39969.1 | |||
BC029015.1 | AAH29015.1 | |||
BC062180.1 | AAH62180.1 | |||
BC094275.1 | AAH94275.1 |
