Assay Details
Target Gene Details
Entrez Gene ID: | 69085 |
Gene Name: | zinc finger, CCHC domain containing 9 |
Gene Aliases: |
1810019C21Rik |
Location: |
Chr.13:91796520-91807997 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc9 | NM_145453.2 | NP_663428.2 | ||
AK007541.1 | BAC25176.1 | |||
AK044802.1 | BAC32099.1 | |||
AK051610.1 | ||||
BB656071.1 |
