Assay Details
Target Gene Details
Entrez Gene ID: | 105518 |
Gene Name: | RIKEN cDNA A630023A22 gene |
Gene Aliases: |
AI461879 |
Location: |
Chr.14:34051126-34102754 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630023A22Rik | NM_001251843.1 | NP_001238772.1 | ||
| AK041587.1 | ||||
| AW989347.1 |
Target Gene Details
Entrez Gene ID: | 11752 |
Gene Name: | annexin A8 |
Gene Aliases: |
AI466995, Anx8 |
Location: |
Chr.14:34085979-34100571 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anxa8 | NM_001281845.1 | NP_001268774.1 | ||
| NM_013473.4 | NP_038501.2 | |||
| AJ002390.1 | CAA05364.1 | |||
| AK009085.1 | ||||
| AK075870.1 | ||||
| BC013271.1 | AAH13271.1 | |||
| BC030407.1 | AAH30407.1 |
More Information
Set Membership: |
|

Genomic Map