Assay Details
Target Gene Details
Entrez Gene ID: | 105243 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 3 |
Gene Aliases: |
9030624O13Rik, AI930210, NHE-3, NHE3 |
Location: |
Chr.13:74086254-74169442 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a3 | NM_001081060.1 | 8 | 1391 | NP_001074529.1 |
More Information
Set Membership: |
|

Genomic Map