Assay Details
Target Gene Details
Entrez Gene ID: | 20504 |
Gene Name: | solute carrier family 17 (sodium phosphate), member 1 |
Gene Aliases: |
NAPI-1, Napi1, Npt1 |
Location: |
Chr.13:23867723-23895730 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a1 | NM_001170638.1 | 3 | 196 | NP_001164109.1 |
| NM_009198.3 | 3 | 288 | NP_033224.3 | |
| NR_121611.1 | 3 | 837 | ||
| AI116499.1 | 3 | 260 | ||
| AI663290.1 | ||||
| AK132800.1 | 3 | 837 | ||
| AK164712.1 | 3 | 180 | ||
| BC013445.1 | 3 | 229 | AAH13445.1 | |
| CJ065843.1 | 3 | 197 | ||
| X77241.1 | 3 | 155 | CAA54459.1 |
More Information
Set Membership: |
|

Genomic Map