Assay Details
Target Gene Details
Entrez Gene ID: | 11792 |
Gene Name: | apurinic/apyrimidinic endonuclease 1 |
Gene Aliases: |
APE, Apex, HAP1, Ref-1 |
Location: |
Chr.14:50924944-50927188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apex1 | NM_009687.2 | NP_033817.1 | ||
AK013296.1 | BAB28774.1 | |||
AK180111.1 | ||||
AK186266.1 | ||||
BC052401.1 | AAH52401.1 | |||
D90374.1 | BAA14382.1 |
