Assay Details
Target Gene Details
Entrez Gene ID: | 11792 |
Gene Name: | apurinic/apyrimidinic endonuclease 1 |
Gene Aliases: |
APE, Apex, HAP1, Ref-1 |
Location: |
Chr.14:50924944-50927188 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apex1 | NM_009687.2 | NP_033817.1 | ||
AK013296.1 | BAB28774.1 | |||
AK180111.1 | ||||
BC052401.1 | AAH52401.1 | |||
D90374.1 | BAA14382.1 |
Target Gene Details
Entrez Gene ID: | 219024 |
Gene Name: | transmembrane protein 55b |
Gene Aliases: |
BC039161 |
Location: |
Chr.14:50926070-50930905 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem55b | NM_001033271.4 | 7 | 2490 | NP_001028443.1 |
AK144251.1 | 7 | 2490 | BAE25798.1 |
