Assay Details
Target Gene Details
Entrez Gene ID: | 66494 |
Gene Name: | PRELI domain containing 1 |
Gene Aliases: |
2610524G07Rik, Preli |
Location: |
Chr.13:55322055-55325272 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prelid1 | NM_025596.5 | NP_079872.4 | ||
AK002846.1 | ||||
AK003629.1 | BAB22898.2 | |||
AK012144.1 | 3 | 618 | ||
AK146857.1 | BAE27486.1 | |||
AK150465.1 | BAE29582.1 | |||
AK151321.1 | BAE30302.1 | |||
AK153455.1 | ||||
AK155042.1 | BAE33009.1 | |||
AK196389.1 | ||||
BC024813.1 | ||||
BC025859.1 | AAH25859.1 | |||
BC098241.1 | AAH98241.1 |
