Assay Details
Target Gene Details
Entrez Gene ID: | 19356 |
Gene Name: | RAD17 homolog (S. pombe) |
Gene Aliases: |
MmRad24 |
Location: |
Chr.13:100617164-100651061 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rad17 | NM_001044371.2 | NP_001037836.1 | ||
NM_001283011.1 | NP_001269940.1 | |||
NM_011233.3 | NP_035363.2 | |||
AF085737.1 | AAC36335.1 | |||
AJ011923.1 | CAA09868.1 | |||
AK134277.1 | BAE22079.1 | |||
AK162016.1 | ||||
AK167358.1 | ||||
BC066855.1 | AAH66855.1 |
