Assay Details
Target Gene Details
Entrez Gene ID: | 66475 |
Gene Name: | ribosomal protein S23 |
Gene Aliases: |
2410044J15Rik |
Location: |
Chr.13:90923122-90924732 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps23 | NM_024175.3 | NP_077137.1 | ||
| AK002573.1 | BAB22198.1 | |||
| AK010594.1 | BAB27050.1 | |||
| AK010608.1 | BAB27058.1 | |||
| AK010667.1 | BAB27102.1 | |||
| AK012287.1 | BAB28145.1 | |||
| AK012438.1 | BAB28238.1 | |||
| AK012574.1 | BAB28327.1 | |||
| AK013720.1 | BAB28969.1 | |||
| AK050580.1 | BAC34329.1 | |||
| AK088085.1 | BAC40136.1 | |||
| AK166994.1 | 4 | 494 | BAE39173.1 | |
| AK168587.1 | BAE40456.1 | |||
| BC002145.1 | AAH02145.1 | |||
| BC054435.1 | AAH54435.1 | |||
| BC078418.1 | AAH78418.1 | |||
| BC100603.1 | AAI00604.1 | |||
| BQ746768.1 |
More Information
Set Membership: |
|

Genomic Map