Assay Details
Target Gene Details
Entrez Gene ID: | 105501 |
Gene Name: | abhydrolase domain containing 4 |
Gene Aliases: |
1110035H23Rik, AI429574, Abh4 |
Location: |
Chr.14:54254129-54269169 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd4 | NM_001205181.1 | NP_001192110.1 | ||
| NM_134076.2 | NP_598837.2 | |||
| AK030250.1 | BAC26863.1 | |||
| AK049366.1 | BAC33711.1 | |||
| AK152595.1 | BAE31344.1 | |||
| AK161853.1 | BAE36606.1 | |||
| AK164655.1 | ||||
| BC017532.1 | AAH17532.1 | |||
| CN713874.1 |

Genomic Map