Assay Details
Target Gene Details
Entrez Gene ID: | 72993 |
Gene Name: | adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1 |
Gene Aliases: |
2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13 |
Location: |
Chr.14:26918986-26971205 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Appl1 | NM_145221.2 | NP_660256.1 | ||
| AK030445.1 | ||||
| AK033566.1 | BAC28364.1 | |||
| AK045438.1 | BAC32367.1 | |||
| AK049307.1 | BAC33672.2 | |||
| AK146356.1 | BAE27108.1 | |||
| AK173169.1 | BAD32447.1 | |||
| AY113705.1 | AAM55531.1 | |||
| BC019708.1 | AAH19708.1 | |||
| BC063751.1 | AAH63751.1 |
More Information
Set Membership: |
|

Genomic Map