Assay Details
Target Gene Details
Entrez Gene ID: | 12177 |
Gene Name: | BCL2/adenovirus E1B interacting protein 3-like |
Gene Aliases: |
C86132, D14Ertd719e, Nip3L, Nix |
Location: |
Chr.14:66985238-67008894 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bnip3l | NM_009761.3 | NP_033891.1 | ||
AF067395.1 | AAD03588.1 | |||
AK004667.1 | BAB23456.1 | |||
AK007920.1 | BAB25351.1 | |||
AK013467.1 | BAB28869.1 | |||
AK018668.1 | BAB31334.1 | |||
AK139950.1 | BAE24194.1 | |||
AK155040.1 | BAE33007.1 | |||
AK168482.1 | BAE40370.1 | |||
AK169391.1 | BAE41138.1 | |||
AY057069.1 | AAL18166.1 | |||
BC085237.1 | AAH85237.1 | |||
BU560379.1 |
