Assay Details
Target Gene Details
Entrez Gene ID: | 319848 |
Gene Name: | solute carrier family 17 (sodium phosphate), member 4 |
Gene Aliases: |
9130214H05Rik |
Location: |
Chr.13:23897879-23915009 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a4 | NM_177016.3 | NP_795990.2 | ||
| AK033669.1 | ||||
| AK050272.1 | BAC34157.1 | |||
| AK162320.1 | ||||
| BC147415.1 | ||||
| BC147416.1 |
More Information
Set Membership: |
|

Genomic Map