Assay Details
Target Gene Details
Entrez Gene ID: | 11843 |
Gene Name: | ADP-ribosylation factor 4 |
Gene Aliases: |
AA407803 |
Location: |
Chr.14:26638197-26657258 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arf4 | NM_007479.3 | 5 | 613 | NP_031505.1 |
| AK002302.1 | 5 | 636 | BAB21999.1 | |
| AK013892.1 | 5 | 668 | BAB29041.1 | |
| AK081686.1 | 5 | 613 | BAC38292.1 | |
| AK153011.1 | 5 | 597 | BAE31650.1 | |
| AK168793.1 | 5 | 634 | BAE40626.1 | |
| AK206427.1 | ||||
| AK208695.1 | 3 | 232 | ||
| AK210624.1 | ||||
| BC115650.1 | 5 | 492 | ||
| BC115651.1 | 5 | 492 | ||
| D87901.1 | 5 | 430 | BAA13493.1 |
More Information
Set Membership: |
|

Genomic Map