Assay Details
Target Gene Details
Entrez Gene ID: | 68808 |
Gene Name: | RIKEN cDNA 1110046J04 gene |
Gene Aliases: |
- |
Location: |
Chr.13:33936032-33960181 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110046J04Rik | NR_040707.1 | |||
| AK004186.1 | ||||
| AK087085.1 | 1 | 477 |
Target Gene Details
Entrez Gene ID: | 20719 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6a |
Gene Aliases: |
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin |
Location: |
Chr.13:33917918-34002794 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb6a | NM_001164117.1 | 1 | 42 | NP_001157589.1 |
| NM_001164118.1 | 1 | 42 | NP_001157590.1 | |
| NM_001243192.1 | NP_001230121.1 | |||
| NM_009254.3 | 1 | 42 | NP_033280.1 | |
| BC006766.1 | AAH06766.1 | |||
| CF732441.1 |
More Information
Set Membership: |
|

Genomic Map